2-Chloro-3-(difluoromethyl)-8-methylquinoline

Lieferant: Apollo Scientific

PC520037-1G
APOSPC520037-1GEA 2000 EUR
APOSPC520037-1G
2-Chloro-3-(difluoromethyl)-8-methylquinoline
2-Chloro-3-(difluoromethyl)-8-methylquinoline
Formel: C₁₁H₈ClF₂N
Molecular Weight: 227,638 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767521
CAS-Nummer: 2090445-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR