3-Bromo-2,4-bis(difluoromethyl)pyridine
Lieferant: Apollo Scientific
PC520038-1G
APOSPC520038-1GEA
4330
EUR
APOSPC520038-1G
3-Bromo-2,4-bis(difluoromethyl)pyridine
3-Bromo-2,4-bis(difluoromethyl)pyridine
Formel:
C₇H₄BrF₄N Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767522 CAS-Nummer: 2091581-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...