tert-Butyl N-[3-(difluoromethyl)-2-pyridyl]carbamate

Lieferant: Apollo Scientific

PC520039-1G
APOSPC520039-1GEA 3190 EUR
APOSPC520039-1G
tert-Butyl N-[3-(difluoromethyl)-2-pyridyl]carbamate
tert-Butyl N-[3-(difluoromethyl)-2-pyridyl]carbamate
Formel: C₁₁H₁₄F₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767523
CAS-Nummer: 1816284-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR