2-Chloro-3-(difluoromethyl)-6-methylquinoline

Lieferant: Apollo Scientific

PC520040-1G
APOSPC520040-1GEA 2270 EUR
APOSPC520040-1G
2-Chloro-3-(difluoromethyl)-6-methylquinoline
2-Chloro-3-(difluoromethyl)-6-methylquinoline
Formel: C₁₁H₈ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD29767524
CAS-Nummer: 2091775-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR