tert-Butyl N-[6-chloro-4-(difluoromethyl)-3-pyridyl]carbamate

Lieferant: Apollo Scientific

PC520042-1G
APOSPC520042-1GEA 3420 EUR
APOSPC520042-1G
tert-Butyl N-[6-chloro-4-(difluoromethyl)-3-pyridyl]carbamate
tert-Butyl N-[6-chloro-4-(difluoromethyl)-3-pyridyl]carbamate
Formel: C₁₁H₁₃ClF₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767525
CAS-Nummer: 2149597-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR