2,4,6-Trichloro-5-(difluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC520044-1G
APOSPC520044-1GEA 1490 EUR
APOSPC520044-1G
2,4,6-Trichloro-5-(difluoromethyl)pyrimidine
2,4,6-Trichloro-5-(difluoromethyl)pyrimidine
Formel: C₅HCl₃F₂N₂
Molecular Weight: 233,431 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767527
CAS-Nummer: 1807617-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR