4,6-Dichloro-5-(difluoromethyl)-2-methyl-pyrimidine
Lieferant: Apollo Scientific
PC520045-1G
APOSPC520045-1GEA
836
EUR
APOSPC520045-1G
4,6-Dichloro-5-(difluoromethyl)-2-methyl-pyrimidine
4,6-Dichloro-5-(difluoromethyl)-2-methyl-pyrimidine
Formel:
C₆H₄Cl₂F₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767528 CAS-Nummer: 2091563-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...