4,6-Dichloro-5-(difluoromethyl)-2-methylsulphanyl-pyrimidine
Lieferant: Apollo Scientific
PC520046-1G
APOSPC520046-1GEA
2280
EUR
APOSPC520046-1G
4,6-Dichloro-5-(difluoromethyl)-2-methylsulphanyl-pyrimidine
4,6-Dichloro-5-(difluoromethyl)-2-methylsulphanyl-pyrimidine
Formel:
C₆H₄Cl₂F₂N₂S Molecular Weight: 245,077 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767529 CAS-Nummer: 2090187-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...