4-Chloro-5-(difluoromethyl)-6-pyrrolidin-1-yl-pyrimidin-2-amine

Lieferant: Apollo Scientific

PC520047-1G
APOSPC520047-1GEA 3650 EUR
APOSPC520047-1G
4-Chloro-5-(difluoromethyl)-6-pyrrolidin-1-yl-pyrimidin-2-amine
4-Chloro-5-(difluoromethyl)-6-pyrrolidin-1-yl-pyrimidin-2-amine
Formel: C₉H₁₁ClF₂N₄
Molecular Weight: 248,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767530
CAS-Nummer: 2091004-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR