4,6-Dichloro-5-(difluoromethyl)pyrimidin-2-amine

Lieferant: Apollo Scientific

PC520048-1G
APOSPC520048-1GEA 1940 EUR
APOSPC520048-1G
4,6-Dichloro-5-(difluoromethyl)pyrimidin-2-amine
4,6-Dichloro-5-(difluoromethyl)pyrimidin-2-amine
Formel: C₅H₃Cl₂F₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD29767531
CAS-Nummer: 2091217-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR