4-Chloro-5-(difluoromethyl)-6-morpholino-pyrimidin-2-amine
Lieferant: Apollo Scientific
PC520049-1G
APOSPC520049-1GEA
3420
EUR
APOSPC520049-1G
4-Chloro-5-(difluoromethyl)-6-morpholino-pyrimidin-2-amine
4-Chloro-5-(difluoromethyl)-6-morpholino-pyrimidin-2-amine
Formel:
C₉H₁₁ClF₂N₄O Molecular Weight: 264,66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767532 CAS-Nummer: 2149598-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...