5-(Difluoromethyl)-2-methylsulphanyl-pyrimidin-4-amine
Lieferant: Apollo Scientific
PC520050-1G
APOSPC520050-1GEA
3420
EUR
APOSPC520050-1G
5-(Difluoromethyl)-2-methylsulphanyl-pyrimidin-4-amine
5-(Difluoromethyl)-2-methylsulphanyl-pyrimidin-4-amine
Formel:
C₆H₇F₂N₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767533 CAS-Nummer: 2090187-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...