4-Chloro-5-(difluoromethyl)-6-(1-piperidyl)pyrimidine

Lieferant: Apollo Scientific

PC520052-1G
APOSPC520052-1GEA 2740 EUR
APOSPC520052-1G
4-Chloro-5-(difluoromethyl)-6-(1-piperidyl)pyrimidine
4-Chloro-5-(difluoromethyl)-6-(1-piperidyl)pyrimidine
Formel: C₁₀H₁₂ClF₂N₃
Molecular Weight: 247,672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767535
CAS-Nummer: 2092708-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR