5-(Difluoromethyl)-2-methoxy-N-methyl-pyrimidin-4-amine
Lieferant: Apollo Scientific
PC520053-1G
APOSPC520053-1GEA
2740
EUR
APOSPC520053-1G
5-(Difluoromethyl)-2-methoxy-N-methyl-pyrimidin-4-amine
5-(Difluoromethyl)-2-methoxy-N-methyl-pyrimidin-4-amine
Formel:
C₇H₉F₂N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767536 CAS-Nummer: 2090446-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...