4,6-Dichloro-5-(difluoromethyl)-2-pyrrolidin-1-yl-pyrimidine

Lieferant: Apollo Scientific

PC520055-1G
APOSPC520055-1GEA 2280 EUR
APOSPC520055-1G
4,6-Dichloro-5-(difluoromethyl)-2-pyrrolidin-1-yl-pyrimidine
4,6-Dichloro-5-(difluoromethyl)-2-pyrrolidin-1-yl-pyrimidine
Formel: C₉H₉Cl₂F₂N₃
Molecular Weight: 268,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767538
CAS-Nummer: 2091581-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR