5-(Difluoromethyl)-2-methoxy-pyrimidine
Lieferant: Apollo Scientific
PC520058-1G
APOSPC520058-1GEA
1490
EUR
APOSPC520058-1G
5-(Difluoromethyl)-2-methoxy-pyrimidine
5-(Difluoromethyl)-2-methoxy-pyrimidine
Formel:
C₆H₆F₂N₂O Molecular Weight: 160,121 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767541 CAS-Nummer: 2128733-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...