5-(Difluoromethyl)-2-methoxy-pyrimidin-4-amine
Lieferant: Apollo Scientific
PC520060-1G
APOSPC520060-1GEA
3420
EUR
APOSPC520060-1G
5-(Difluoromethyl)-2-methoxy-pyrimidin-4-amine
5-(Difluoromethyl)-2-methoxy-pyrimidin-4-amine
Formel:
C₆H₇F₂N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767543 CAS-Nummer: 2092285-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...