5-(Difluoromethyl)-2-pyrrolidin-1-yl-pyrimidine
Lieferant: Apollo Scientific
PC520061-1G
APOSPC520061-1GEA
1490
EUR
APOSPC520061-1G
5-(Difluoromethyl)-2-pyrrolidin-1-yl-pyrimidine
5-(Difluoromethyl)-2-pyrrolidin-1-yl-pyrimidine
Formel:
C₉H₁₁F₂N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767544 CAS-Nummer: 2128733-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...