tert-Butyl 4-[6-chloro-5-(difluoromethyl)pyrimidin-4-yl]piperazine-1-carboxylate

Lieferant: Apollo Scientific

PC520062-1G
APOSPC520062-1GEA 2740 EUR
APOSPC520062-1G
tert-Butyl 4-[6-chloro-5-(difluoromethyl)pyrimidin-4-yl]piperazine-1-carboxylate
tert-Butyl 4-[6-chloro-5-(difluoromethyl)pyrimidin-4-yl]piperazine-1-carboxylate
Formel: C₁₄H₁₉ClF₂N₄O₂
Molecular Weight: 348.776 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767545
CAS-Nummer: 2149590-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR