5-(Difluoromethyl)-2,4-dimethoxy-pyrimidine

Lieferant: Apollo Scientific

PC520063-1G
APOSPC520063-1GEA 1550 EUR
APOSPC520063-1G
5-(Difluoromethyl)-2,4-dimethoxy-pyrimidine
5-(Difluoromethyl)-2,4-dimethoxy-pyrimidine
Formel: C₇H₈F₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767546
CAS-Nummer: 2128733-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR