Methyl 2-(difluoromethyl)pyrimidine-5-carboxylate
Lieferant: Apollo Scientific
PC520065-1G
APOSPC520065-1GEA
3420
EUR
APOSPC520065-1G
Methyl 2-(difluoromethyl)pyrimidine-5-carboxylate
Methyl 2-(difluoromethyl)pyrimidine-5-carboxylate
Formel:
C₇H₆F₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767548 CAS-Nummer: 2120555-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...