6-Chloro-5-(difluoromethyl)pyrimidin-4-amine

Lieferant: Apollo Scientific

PC520067-1G
APOSPC520067-1GEA 2280 EUR
APOSPC520067-1G
6-Chloro-5-(difluoromethyl)pyrimidin-4-amine
6-Chloro-5-(difluoromethyl)pyrimidin-4-amine
Formel: C₅H₄ClF₂N₃
Molecular Weight: 179,555 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767551
CAS-Nummer: 2092036-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR