5-Bromo-3-(difluoromethyl)pyrazin-2-amine

Lieferant: Apollo Scientific

PC520068-1G
APOSPC520068-1GEA 3650 EUR
APOSPC520068-1G
5-Bromo-3-(difluoromethyl)pyrazin-2-amine
5-Bromo-3-(difluoromethyl)pyrazin-2-amine
Formel: C₅H₄BrF₂N₃
Molecular Weight: 224,006 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767552
CAS-Nummer: 2092522-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR