2-(Difluoromethyl)-5-methoxy-3-methyl-1H-indole

Lieferant: Apollo Scientific

PC520071-1G
APOSPC520071-1GEA 797 EUR
APOSPC520071-1G
2-(Difluoromethyl)-5-methoxy-3-methyl-1H-indole
2-(Difluoromethyl)-5-methoxy-3-methyl-1H-indole
Formel: C₁₁H₁₁F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD29767555
CAS-Nummer: 2149590-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR