6-Bromo-2-(difluoromethyl)-1H-indole
Lieferant: Apollo Scientific
|
Gefahr
|
PC520072-1G
APOSPC520072-1GEA
2280
EUR
APOSPC520072-1G
6-Bromo-2-(difluoromethyl)-1H-indole
6-Bromo-2-(difluoromethyl)-1H-indole
Formel:
C₉H₆BrF₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767556 CAS-Nummer: 1780771-02-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...