4-benzyloxy-3-(difluoromethyl)-1H-indole

Lieferant: Apollo Scientific

PC520074-1G
APOSPC520074-1GEA 1550 EUR
APOSPC520074-1G
4-benzyloxy-3-(difluoromethyl)-1H-indole
4-benzyloxy-3-(difluoromethyl)-1H-indole
Formel: C₁₆H₁₃F₂NO
Molecular Weight: 273,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767558
CAS-Nummer: 2149598-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR