3-(Difluoromethyl)-1H-indole-5-carbonitrile

Lieferant: Apollo Scientific

PC520076-1G
APOSPC520076-1GEA 1550 EUR
APOSPC520076-1G
3-(Difluoromethyl)-1H-indole-5-carbonitrile
3-(Difluoromethyl)-1H-indole-5-carbonitrile
Formel: C₁₀H₆F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767560
CAS-Nummer: 2114943-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR