7-benzyloxy-3-(difluoromethyl)-1H-indole

Lieferant: Apollo Scientific

PC520080-1G
APOSPC520080-1GEA 1490 EUR
APOSPC520080-1G
7-benzyloxy-3-(difluoromethyl)-1H-indole
7-benzyloxy-3-(difluoromethyl)-1H-indole
Formel: C₁₆H₁₃F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD29767564
CAS-Nummer: 2149590-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR