7-Bromo-3-(difluoromethyl)-1H-indole
Lieferant: Apollo Scientific
PC520086-1G
APOSPC520086-1GEA
1710
EUR
APOSPC520086-1G
7-Bromo-3-(difluoromethyl)-1H-indole
7-Bromo-3-(difluoromethyl)-1H-indole
Formel:
C₉H₆BrF₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767570 CAS-Nummer: 1780756-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...