2-Bromo-3-(difluoromethyl)-1H-indole

Lieferant: Apollo Scientific

PC520087-1G
APOSPC520087-1GEA 2280 EUR
APOSPC520087-1G
2-Bromo-3-(difluoromethyl)-1H-indole
2-Bromo-3-(difluoromethyl)-1H-indole
Formel: C₉H₆BrF₂N
Molecular Weight: 246.051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767571
CAS-Nummer: 2092048-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR