6-benzyloxy-3-(difluoromethyl)-1H-indole

Lieferant: Apollo Scientific

PC520088-1G
APOSPC520088-1GEA 1710 EUR
APOSPC520088-1G
6-benzyloxy-3-(difluoromethyl)-1H-indole
6-benzyloxy-3-(difluoromethyl)-1H-indole
Formel: C₁₆H₁₃F₂NO
Molecular Weight: 273,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767572
CAS-Nummer: 2149598-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR