6-Bromo-3-(difluoromethyl)-N,N-dimethyl-1H-indol-2-amine
Lieferant: Apollo Scientific
PC520090-1G
APOSPC520090-1GEA
3190
EUR
APOSPC520090-1G
6-Bromo-3-(difluoromethyl)-N,N-dimethyl-1H-indol-2-amine
6-Bromo-3-(difluoromethyl)-N,N-dimethyl-1H-indol-2-amine
Formel:
C₁₁H₁₁BrF₂N₂ Molecular Weight: 289,119 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767574 CAS-Nummer: 2091011-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...