5-Bromo-3-(difluoromethyl)-1H-indol-4-ol

Lieferant: Apollo Scientific

PC520094-1G
APOSPC520094-1GEA 4530 EUR
APOSPC520094-1G
5-Bromo-3-(difluoromethyl)-1H-indol-4-ol
5-Bromo-3-(difluoromethyl)-1H-indol-4-ol
Formel: C₉H₆BrF₂NO
Molecular Weight: 262,051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767578
CAS-Nummer: 2091581-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR