[3-(Difluoromethyl)-5-methoxy-1H-indol-4-yl]azinate

Lieferant: Apollo Scientific

PC520096-1G
APOSPC520096-1GEA 3650 EUR
APOSPC520096-1G
[3-(Difluoromethyl)-5-methoxy-1H-indol-4-yl]azinate
[3-(Difluoromethyl)-5-methoxy-1H-indol-4-yl]azinate
Formel: C₁₀H₈F₂N₂O₃
Molecular Weight: 242,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496607
CAS-Nummer: 2149600-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR