3-(Difluoromethyl)-2-(2,5-dimethoxyphenyl)-1H-indole

Lieferant: Apollo Scientific

PC520102-1G
APOSPC520102-1GEA 3650 EUR
APOSPC520102-1G
3-(Difluoromethyl)-2-(2,5-dimethoxyphenyl)-1H-indole
3-(Difluoromethyl)-2-(2,5-dimethoxyphenyl)-1H-indole
Formel: C₁₇H₁₅F₂NO₂
Molecular Weight: 303,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767585
CAS-Nummer: 2149597-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR