Methyl 3-(difluoromethyl)-1H-indole-5-carboxylate

Lieferant: Apollo Scientific

PC520104-1G
APOSPC520104-1GEA 2280 EUR
APOSPC520104-1G
Methyl 3-(difluoromethyl)-1H-indole-5-carboxylate
Methyl 3-(difluoromethyl)-1H-indole-5-carboxylate
Formel: C₁₁H₉F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767587
CAS-Nummer: 2112530-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR