4-Bromo-3-(difluoromethyl)-1H-indole

Lieferant: Apollo Scientific

PC520107-1G
APOSPC520107-1GEA 1550 EUR
APOSPC520107-1G
4-Bromo-3-(difluoromethyl)-1H-indole
4-Bromo-3-(difluoromethyl)-1H-indole
Formel: C₉H₆BrF₂N
Molecular Weight: 246,051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767590
CAS-Nummer: 1780756-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR