3-Bromo-6-(difluoromethyl)-1H-indole
Lieferant: Apollo Scientific
PC520116-1G
APOSPC520116-1GEA
3650
EUR
APOSPC520116-1G
3-Bromo-6-(difluoromethyl)-1H-indole
3-Bromo-6-(difluoromethyl)-1H-indole
Formel:
C₉H₆BrF₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767599 CAS-Nummer: 2091766-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...