5-Bromo-4-chloro-2-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

PC520120-1G
APOSPC520120-1GEA 2740 EUR
APOSPC520120-1G
5-Bromo-4-chloro-2-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₄BrClF₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767603
CAS-Nummer: 2092583-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR