2-(Difluoromethyl)-5-fluoro-6-methoxy-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

PC520122-1G
APOSPC520122-1GEA 4540 EUR
APOSPC520122-1G
2-(Difluoromethyl)-5-fluoro-6-methoxy-1H-pyrrolo[2,3-b]pyridine
2-(Difluoromethyl)-5-fluoro-6-methoxy-1H-pyrrolo[2,3-b]pyridine
Formel: C₉H₇F₃N₂O
Molecular Weight: 216.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767605
CAS-Nummer: 2149590-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR