4-Chloro-2-(difluoromethyl)-5-fluoro-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

PC520123-1G
APOSPC520123-1GEA 3650 EUR
APOSPC520123-1G
4-Chloro-2-(difluoromethyl)-5-fluoro-1H-pyrrolo[2,3-b]pyridine
4-Chloro-2-(difluoromethyl)-5-fluoro-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₄ClF₃N₂
Molecular Weight: 220,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767606
CAS-Nummer: 2092048-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR