4-Chloro-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Lieferant: Apollo Scientific
PC520124-1G
APOSPC520124-1GEA
2280
EUR
APOSPC520124-1G
4-Chloro-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
4-Chloro-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel:
C₈H₅ClF₂N₂ Molecular Weight: 202,589 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767607 CAS-Nummer: 2092851-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...