6-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

PC520125-1G
APOSPC520125-1GEA 2280 EUR
APOSPC520125-1G
6-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
6-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₅BrF₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767608
CAS-Nummer: 2090237-47-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR