3-(Difluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-amine

Lieferant: Apollo Scientific

PC520126-1G
APOSPC520126-1GEA 2280 EUR
APOSPC520126-1G
3-(Difluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-amine
3-(Difluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-amine
Formel: C₈H₇F₂N₃
Molecular Weight: 183,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767609
CAS-Nummer: 2091580-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR