5-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

PC520127-1G
APOSPC520127-1GEA 2280 EUR
APOSPC520127-1G
5-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
5-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₅BrF₂N₂
Molecular Weight: 247,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767610
CAS-Nummer: 2092285-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR