3-Bromo-5-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

PC520128-1G
APOSPC520128-1GEA 2740 EUR
APOSPC520128-1G
3-Bromo-5-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
3-Bromo-5-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₅BrF₂N₂
Molecular Weight: 247,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767611
CAS-Nummer: 2092406-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR