4-Bromo-2-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine

Lieferant: Apollo Scientific

PC520130-1G
APOSPC520130-1GEA 2740 EUR
APOSPC520130-1G
4-Bromo-2-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
4-Bromo-2-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
Formel: C₈H₅BrF₂N₂
Molecular Weight: 247,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767613
CAS-Nummer: 2091580-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR