5-Chloro-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine

Lieferant: Apollo Scientific

PC520131-1G
APOSPC520131-1GEA 2740 EUR
APOSPC520131-1G
5-Chloro-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
5-Chloro-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
Formel: C₈H₅ClF₂N₂
Molecular Weight: 202,589 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767614
CAS-Nummer: 2091100-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR