5-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine

Lieferant: Apollo Scientific

PC520132-1G
APOSPC520132-1GEA 2740 EUR
APOSPC520132-1G
5-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
5-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
Formel: C₈H₅BrF₂N₂
Molecular Weight: 247.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767615
CAS-Nummer: 2090237-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR