3-(Difluoromethyl)-1H-pyrrolo[2,3-c]pyridin-4-amine
Lieferant: Apollo Scientific
PC520133-1G
APOSPC520133-1GEA
3650
EUR
APOSPC520133-1G
3-(Difluoromethyl)-1H-pyrrolo[2,3-c]pyridin-4-amine
3-(Difluoromethyl)-1H-pyrrolo[2,3-c]pyridin-4-amine
Formel:
C₈H₇F₂N₃ Molecular Weight: 183,158 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767616 CAS-Nummer: 2091011-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...